Learn More
N-Methoxy-N,2,5,9-tetramethyldeca-2,4,8-trienamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methoxy-N,2,5,9-tetramethyldeca-2,4,8-trienamide |
| CAS | 2508008-05-5 |
| Molecular Formula | C15H25NO2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C15H25NO2/c1-12(2)8-7-9-13(3)10-11-14(4)15(17)16(5)18-6/h8,10-11H,7,9H2,1-6H3 |
| SMILES | C/C(C)=C\CCC(C)=CC=C(C)C(N(OC)C)=O |
| IUPAC Name | N-methoxy-N,2,5,9-tetramethyldeca-2,4,8-trienamide |
| Molecular Weight (g/mol) | 251.36 |
| Formula Weight | 251.19 |