Learn More
N-Isopropyl-N-methylamine-d7 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-Isopropyl-N-methylamine-d7 Hydrochloride |
| CAS | 2733969-46-3 |
| Molecular Formula | C4H4D7N . (HCl) |
| Quantity | 100 mg |
| Synonym | N-Methylisopropylamine-d7 Hydrochloride, Methylisopropylammonium-d7 Chloride |
| InChI Formula | InChI=1S/C4H11N.ClH/c1-4(2)5-3;/h4-5H,1-3H3;1H/i1D3,2D3,4D; |
| SMILES | CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H].Cl |
| IUPAC Name | N-methylpropan-d7-2-amine hydrochloride |
| Molecular Weight (g/mol) | 116.64 |
| Formula Weight | 116.11 |