Learn More
n-Hexane-1,1,1,2,2,3,3-d7, CDN
Product Code. 30323793
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323793 0.05 g -
30328160 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323793 Supplier CDN Supplier No. CDND73300.05G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material n-Hexane-1,1,1,2,2,3,3-D7
CAS 32740-31-1
Molecular Formula C6 D7 H7
Quantity 0.05 g
Synonym Hexane-1,1,1,2,2,3,3-d7 (8 CI, 9 CI), n-Hexane-1,1,1,2,2,3,3-d7, n-Hexane-1,1,1,2,2,3,3-D7, [1,1,1,2,2,3,3-2H7]Hexane
InChI Formula InChI=1 S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3/i1D3,3D2,5D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCC
IUPAC Name 1,1,1,2,2,3,3-heptadeuteriohexane
Molecular Weight (g/mol) 93.219
Formula Weight 93.153 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity