Learn More
N-Formylindoline, TRC
Specifications
Specifications
| Chemical Name or Material | N-Formylindoline |
| CAS | 2861-59-8 |
| Molecular Formula | C9H9NO |
| Quantity | 50 mg |
| Synonym | 2,3-Dihydro-1H-indole-1-carboxaldehyde, 1-Indolinecarboxaldehyde |
| InChI Formula | InChI=1S/C9H9NO/c11-7-10-6-5-8-3-1-2-4-9(8)10/h1-4,7H,5-6H2 |
| SMILES | O=CN1CCc2ccccc12 |
| Molecular Weight (g/mol) | 147.17 |
| Formula Weight | 147.0684 |