Learn More
N-Formylglycine ethyl ester, 97%, Thermo Scientific™
Product Code. 11328435
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328435 25 g 25g
11318435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11328435 Supplier Thermo Scientific Alfa Aesar Supplier No. L10437.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
N-Formylglycine ethyl ester is used in the preparation of aminomethylene compounds. Ethyl isocyanoacetate, sodium enolate was preparedm from ethyl N-formylglycinate.
Solubility
Soluble in common organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3154-51-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00014125
InChI Key GMBCCEOJUWMBPF-UHFFFAOYSA-N
Synonym n-formylglycine ethyl ester, ethyl n-formylglycinate, glycine, n-formyl-, ethyl ester, ethyl formylaminoacetate, ethylformylglycine, n-formylglycineethylester, ethyl formamidoacetate, 2-formylaminoacetic acid ethyl ester, for-gly-oet, ethyl formylglycinate
PubChem CID 76615
IUPAC Name ethyl 2-formamidoacetate
SMILES CCOC(=O)CNC=O
Specifications
Boiling Point 106°C to 107°C (0.5mmHg)
Linear Formula OHCNHCH2CO2CH2CH3
Refractive Index 1.453
Beilstein 1099378
Solubility Information Soluble in common organic solvents.
Density 1.15
Quantity 25 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material N-Formylglycine ethyl ester