Learn More
MedChemExpress N-Formyl-Nle-Leu-Phe-Nle-Tyr-Lys
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Purity | 98.64% |
| CAS | 71901-21-8 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 824.02 g/mol |
| Product Type | Peptides |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 16.67 mg/mL (20.23 mM∣ultrasonic and adjust pH to 12 with NaOH) |
| Synonym | For-Nle-Leu-Phe-Nle-Tyr-Lys-OH |