Learn More
N-Formyl-L-leucine (3S,4R,6S)-3-Hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl Ester, TRC
Product Code. 30007573
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007573 25 mg 25mg
30010954 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30007573 Supplier Toronto Research Chemicals Supplier No. TRCF70057025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 130793-27-0
Molecular Formula C29 H53 N O5
Molecular Weight (g/mol) 495.73
Synonym L-Leucine, N-formyl-, (3S,4R,6S)-3-hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl ester, L-Leucine, N-formyl-, 3-hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl ester, [3S-(3α,4β,6β)]-, Orlistat Related Compound D (USP)
IUPAC Name [(3S,4R,6S)-3-hexyl-2-oxo-6-undecyloxan-4-yl] (2S)-2-formamido-4-methylpentanoate
SMILES CCCCCCCCCCC[C@H]1C[C@@H](OC(=O)[C@H](CC(C)C)NC=O)[C@H](CCCCCC)C(=O)O1
Specifications
InChI Formula InChI=1S/C29H53NO5/c1-5-7-9-11-12-13-14-15-16-18-24-21-27(25(28(32)34-24)19-17-10-8-6-2)35-29(33)26(30-22-31)20-23(3)4/h22-27H,5-21H2,1-4H3,(H,30,31)/t24-,25-,26-,27+/m0/s1
Quantity 25 mg
Formula Weight 495.3924
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-formyl-L-leucine (3S,4R,6S)-3-hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl ester
Grade HPLC