Learn More
N-Fmoc-L-serine, 97+%
Product Code. 11404537
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404537 1 g 1g
11414537 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11404537 Supplier Thermo Scientific Alfa Aesar Supplier No. B21079.03
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 73724-45-5 | C18H16NO5 | 326.33 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73724-45-5
Molecular Formula C18H16NO5
Molecular Weight (g/mol) 326.33
MDL Number MFCD00051928
InChI Key JZTKZVJMSCONAK-INIZCTEOSA-M
Synonym fmoc-ser-oh, fmoc-l-serine, fmoc-l-ser-oh, n-fmoc-l-serine, n-9-fluorenylmethoxycarbonyl-l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-l-serine, n-9h-fluoren-9-ylmethoxycarbonyl serine, s-fmoc-2-amino-3-hydroxypropionic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-serine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid
PubChem CID 6541433
SMILES OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
Specifications
Melting Point 102°C to 106°C
Quantity 1 g
Beilstein 4715791
Optical Rotation +13.6° (c=1 in Ethyl Acetate)
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxypropanoic acid
Formula Weight 327.34
Percent Purity ≥97%
Chemical Name or Material N-Fmoc-L-serine