missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-pentafluorophenylalanine, 95%

Product Code. 15428567
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428567 250 mg 250mg
15408567 1 g 1g
15418567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428567 Supplier Thermo Scientific Alfa Aesar Supplier No. H52059.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 205526-32-5 | C24H16F5NO4 | 477.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205526-32-5
Molecular Formula C24H16F5NO4
Molecular Weight (g/mol) 477.39
MDL Number MFCD00270612
InChI Key DLOGILOIJKBYKA-KRWDZBQOSA-N
Synonym fmoc-pentafluoro-l-phenylalanine, fmoc-phe f5-oh, fmoc-l-pentafluorophenylalanine, fmoc-l-pentafluorophe, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-perfluorophenyl propanoic acid, fmoc-3-pentafluorophenyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-pentafluoro-l-phenylalanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-pentafluoro-phenylalanine, s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pentafluorophenyl-propionic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pentafluorophenyl propanoic acid
PubChem CID 7020337
SMILES OC(=O)[C@H](CC1=C(F)C(F)=C(F)C(F)=C1F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 250 mg
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
Formula Weight 477.39
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-pentafluorophenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.