Learn More
N-Despropyl (R)-Ropivacaine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Despropyl (R)-Ropivacaine |
| CAS | 27262-43-7 |
| Molecular Formula | C14 H20 N2 O |
| Quantity | 100 mg |
| Synonym | (2R)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide, (R)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide, (R)-2',6'-Pipecoloxylidide, (-)-2',6'-Pipecoloxylidide, (-)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide, N-Desbutyl (R)-Bupivacaine, N-Desmethyl (R)-Mepivacaine |
| InChI Formula | InChI=1S/C14H20N2O/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12/h5-7,12,15H,3-4,8-9H2,1-2H3,(H,16,17)/t12-/m1/s1 |
| SMILES | Cc1cccc(C)c1NC(=O)[C@H]2CCCCN2 |
| IUPAC Name | (2R)-N-(2,6-dimethylphenyl)piperidine-2-carboxamide |
| Molecular Weight (g/mol) | 232.321 |
| Formula Weight | 232.158 |