missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethylclozapine, MedChemExpress

Product Code. p-200098859
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16498798

Brand: Medchem Express HYG0021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-Desmethylclozapine is a major active metabolite of the atypical antipsychotic drug Clozapine. N-Desmethylclozapine is a potent, allosteric and partial M1 receptors agonist (EC50=115 nM) and is able to potentiate hippocampal N-methyl-d-aspartate (NMDA) receptor currents through M1 receptor activation. N-Desmethylclozapine is also a δ-opioid agonist.

N-Desmethylclozapine is a major active metabolite of the atypical antipsychotic drug Clozapine. N-Desmethylclozapine is a potent, allosteric and partial M1 receptors agonist (EC50=115 nM) and is able to potentiate hippocampal N-methyl-d-aspartate (NMDA) receptor currents through M1 receptor activation. N-Desmethylclozapine is also a δ-opioid agonist.

Specifications

Chemical Name or Material N-Desmethylclozapine
CAS 6104-71-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H17ClN4
Quantity 10 mg
Synonym Norclozapine Desmethylclozapine Normethylclozapine
Solubility Information DMSO : ≥ 50 mg/mL (159.85 mM)
SMILES ClC1=CC=C2C(N=C(N3CCNCC3)C4=CC=CC=C4N2)=C1
Molecular Weight (g/mol) 312.8
Formula Weight 312.8
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.