Learn More
N-Desmethyl Sorafenib, TRC
Specifications
Specifications
| Chemical Name or Material | N-Desmethylsorafenib |
| CAS | 284461-74-1 |
| Molecular Formula | C21H15ClF3N3O3 |
| Quantity | 1 g |
| Synonym | 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-2-pyridinecarboxamide, BAY 43-9007, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N’-[4-(2-carbamoyl-4-pyridyloxy)phenyl]urea |
| InChI Formula | InChI=1S/C21H15ClF3N3O3/c22-18-9-6-14(11-17(18)21(23,24)25)28-20(30)27-13-4-7-15(8-5-13)31-16-3-1-2-12(10-16)19(26)29/h1-11H,(H2,26,29)(H2,27,28,30) |
| SMILES | NC(=O)c1cccc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)cc2)c1 |
| IUPAC Name | 3-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]benzamide |
| Molecular Weight (g/mol) | 449.81 |
| Formula Weight | 449.075 |