Learn More
N-Desmethyl N-Nitroso Imatinib, TRC
Specifications
Specifications
| Chemical Name or Material | N-Desmethyl N-Nitroso Imatinib |
| CAS | 2816369-80-7 |
| Molecular Formula | C28 H28 N8 O2 |
| Quantity | 50 mg |
| Synonym | N-(4-Methyl-3((4-(pyridin-3yl)pyrimidin-2-yl)amino)phenyl)-4-((4-nitrosopiperazin-1-yl)methyl)benzamide |
| InChI Formula | InChI=1S/C28H28N8O2/c1-20-4-9-24(17-26(20)33-28-30-12-10-25(32-28)23-3-2-11-29-18-23)31-27(37)22-7-5-21(6-8-22)19-35-13-15-36(34-38)16-14-35/h2-12,17-18H,13-16,19H2,1H3,(H,31,37)(H,30,32,33) |
| SMILES | Cc1ccc(NC(=O)c2ccc(CN3CCN(CC3)N=O)cc2)cc1Nc4nccc(n4)c5cccnc5 |
| IUPAC Name | N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]-4-[(4-nitrosopiperazin-1-yl)methyl]benzamide |
| Molecular Weight (g/mol) | 508.574 |
| Formula Weight | 508.234 |