Learn More
N-Desmethyl-benzotropine Hydrochloride, TRC
Product Code. 30363993
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363993 25 mg 25mg
30364293 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30363993 Supplier Toronto Research Chemicals Supplier No. TRCD29104025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25471-67-4
Molecular Formula C20 H23 N O . Cl H
Molecular Weight (g/mol) 329.86
Synonym 8-Azabicyclo[3.2.1]octane, 3-(diphenylmethoxy)-, hydrochloride, endo- (9CI), 1αH,5αH-Nortropane, 3α-(diphenylmethoxy)-, hydrochloride (8CI), (3-endo)-3-(Diphenylmethoxy)-8-azabicyclo[3.2.1]octane hydrochloride, endo-3-(Benzhydryloxy)-8-azabicyclo[3.2.1]octane hydrochloride, Desmethyl benzatropine hydrochloride, N-Desmethylbenzatropine hydrochloride, N-Desmethylbenztropine hydrochloride
IUPAC Name (1R,5S)-3-benzhydryloxy-8-azabicyclo[3.2.1]octane;hydrochloride
SMILES Cl.C1C[C@H]2C[C@H](C[C@@H]1N2)OC(c3ccccc3)c4ccccc4
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C20H23NO.ClH/c1-3-7-15(8-4-1)20(16-9-5-2-6-10-16)22-19-13-17-11-12-18(14-19)21-17;/h1-10,17-21H,11-14H2;1H/t17-,18+,19+;
Formula Weight 329.1546
Chemical Name or Material N-Desmethyl Benzotropine hydrochloride