Learn More
N-Desbutyl-N-propyl Bumetanide, TRC
Specifications
Specifications
| Chemical Name or Material | N-Desbutyl-N-propyl Bumetanide |
| CAS | 28395-28-0 |
| Molecular Formula | C16 H18 N2 O5 S |
| Quantity | 50 mg |
| Synonym | 3-(Aminosulfonyl)-4-phenoxy-5-(propylamino)benzoic Acid, 4-Phenoxy-3-(propylamino)-5-sulfamoylbenzoic Acid, 4-Phenoxy-3-propylamino-5-sulfamoylbenzoic Acid |
| InChI Formula | InChI=1S/C16H18N2O5S/c1-2-8-18-13-9-11(16(19)20)10-14(24(17,21)22)15(13)23-12-6-4-3-5-7-12/h3-7,9-10,18H,2,8H2,1H3,(H,19,20)(H2,17,21,22) |
| SMILES | CCCNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O |
| IUPAC Name | 4-phenoxy-3-(propylamino)-5-sulfamoylbenzoic acid |
| Molecular Weight (g/mol) | 350.389 |
| Formula Weight | 350.094 |