Learn More
N-des-succinyl-N-Boc-O-desethyl Sacubitril, TRC
Specifications
Specifications
| Chemical Name or Material | (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic Acid |
| CAS | 1012341-50-2 |
| Molecular Formula | C23H29NO4 |
| Quantity | 500 mg |
| Synonym | (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic Acid, (alphaR,gammaS)-gamma-[[(1,1-Dimethylethoxy)carbonyl]amino]-alpha-methyl[1,1'-biphenyl]-4-pentanoic Acid |
| InChI Formula | InChI=1S/C23H29NO4/c1-16(21(25)26)14-20(24-22(27)28-23(2,3)4)15-17-10-12-19(13-11-17)18-8-6-5-7-9-18/h5-13,16,20H,14-15H2,1-4H3,(H,24,27)(H,25,26)/t16-,20+/m1/s1 |
| SMILES | C[C@H](C[C@@H](Cc1ccc(cc1)c2ccccc2)NC(=O)OC(C)(C)C)C(=O)O |
| IUPAC Name | (2R,4S)-2-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(4-phenylphenyl)pentanoic acid |
| Molecular Weight (g/mol) | 383.48 |
| Formula Weight | 383.2097 |