Learn More
N-Demethyl Roxithromycin, TRC
Product Code. 30012443
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012443 25 mg 25mg
17981905 5 mg 5mg
17994945 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012443 Supplier Toronto Research Chemicals Supplier No. TRCD23126525MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118267-18-8
Molecular Formula C40H74N2O15
Molecular Weight (g/mol) 823.02
Synonym Roxithromycin Ep Impurity F
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@H](NC)[C@H]3O)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)\[C@H](C)[C@@H](O)[C@]1(C)O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C40H74N2O15/c1-14-28-40(10,48)33(44)23(4)30(42-52-20-51-16-15-49-12)21(2)18-38(8,47)35(57-37-31(43)27(41-11)17-22(3)53-37)24(5)32(25(6)36(46)55-28)56-29-19-39(9,50-13)34(45)26(7)54-29/h21-29,31-35,37,41,43-45,47-48H,14-20H2,1-13H3/b42-30-/t21-,22-,23+,24+,25-,26+,27+,28-,29+,31-,32+,33-,34+,35-,37+,38-,39-,40-/m1/s1
Formula Weight 822.5089
Percent Purity >95%
Chemical Name or Material N-Demethyl Roxithromycin
Grade HPLC