Learn More
n-Decyl-1,1,2,2-d4 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Decanol-1,1,2,2-D4 |
| CAS | 57367-97-2 |
| Molecular Formula | C10H18D4O |
| Quantity | 0.5 g |
| Synonym | 1-Decan-1,1,2,2-d4-ol (9 CI), 1,1,2,2-Tetradeuteriodecan-1-ol, 1-Decanol-1,1,2,2-d4, 1-Decanol-1,1,2,2-D4, n-Decyl-1,1,2,2-d4 alcohol |
| InChI Formula | InChI=1 S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3/i9D2,10D2 |
| SMILES | CCCCCCCCC([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | decan-1,1,2,2-d4-1-ol |
| Molecular Weight (g/mol) | 162.31 |
| Formula Weight | 162.19 g/mol |
| Show More |