Learn More
N-Chloroacetyl-tert-butylamine, TRC
Product Code. 30350436
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350436 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30350436 Supplier Toronto Research Chemicals Supplier No. TRCC3633902.5G
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Chloroacetyl-tert-butylamine
CAS 15678-99-6
Molecular Formula C6H12ClNO
Quantity 2.5 g
Synonym alpha-Chloro-N-tert-butylacetamide, 2-Chloro-N-t-butylacetamide, N-Chloroacetyl-tert-butylamine, N-tert-Butyl-2-chloroacetamide, NSC 8361
InChI Formula InChI=1S/C6H12ClNO/c1-6(2,3)8-5(9)4-7/h4H2,1-3H3,(H,8,9)
SMILES CC(C)(C)NC(=O)CCl
IUPAC Name N-tert-butyl-2-chloroacetamide
Molecular Weight (g/mol) 149.06
Formula Weight 149.0607