Learn More
N-(Carboxypropyl)-N,N,N-trimethyl-d9-ammonium Chloride, CDN
Product Code. 30321604
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321604 0.05 g 0.05g
30318105 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321604 Supplier CDN Supplier No. CDND67890.05G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material gamma-Butyrobetaine trimethyl-d9 hydrochloride
CAS 85806-17-3
Molecular Formula C7 2H9 H7 N O2 . Cl
Quantity 0.05 g
InChI Formula InChI=1 S/C7H15NO2.ClH/c1-8(2,3)6-4-5-7(9)10;/h4-6H2,1-3H3;1 H/i1D3,2D3,3D3;
SMILES [Cl-].[2 H]C([2 H])([2 H])[N+](CCCC(=O)O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 3-carboxypropyl-tris(trideuteriomethyl)azanium;chloride
Molecular Weight (g/mol) 190.72
Formula Weight 190.1434 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity