Learn More
N-(Carboxypropyl)-N,N,N-trimethyl-d9-ammonium Chloride, CDN
Specifications
Specifications
| Chemical Name or Material | gamma-Butyrobetaine trimethyl-d9 hydrochloride |
| CAS | 85806-17-3 |
| Molecular Formula | C7 2H9 H7 N O2 . Cl |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C7H15NO2.ClH/c1-8(2,3)6-4-5-7(9)10;/h4-6H2,1-3H3;1 H/i1D3,2D3,3D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](CCCC(=O)O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-carboxypropyl-tris(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 190.72 |
| Formula Weight | 190.1434 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |