Learn More
N-Carbamoylmaleimide, tech. 90%
Product Code. 11434654
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434654 10 g 10g
11444654 50 g 50g
11454654 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11434654 Supplier Thermo Scientific Alfa Aesar Supplier No. A13685.09
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 3345-50-4 | C5H4N2O3 | 140.098 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3345-50-4
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00014539
InChI Key BNPFHEFZJPVCCE-UHFFFAOYSA-N
Synonym n-carbamoylmaleimide, n-carbamylmaleimide, 2,5-dioxo-3-pyrroline-1-carboxamide, 2,5-dioxo-2,5-dihydro-1h-pyrrole-1-carboxamide, n-carbamoylmaleiimide, 3-pyrrolinecarboxamide, 2,5-dioxo, 3-pyrroline-2,5-dione, 1-carbamoyl, 1h-pyrrole-1-carboxamide, 2,5-dihydro-2,5-dioxo-9ci, 2,5-dihydro-2,5-dioxo-1h-pyrrole-1-carboxamide, 2,5-dioxoazolinecarboxamide
PubChem CID 76868
IUPAC Name 2,5-dioxopyrrole-1-carboxamide
SMILES C1=CC(=O)N(C1=O)C(=O)N
Specifications
Melting Point 157°C to 160°C
Quantity 10 g
Beilstein 117919
Formula Weight 140.1
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Carbamoylmaleimide