Learn More
n-Butylbenzene, 99+%
Product Code. p-79168 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
Product Code. 10530171
Brand: Thermo Scientific Acros 107850500
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 104-51-8 | C10H14 | 134.22 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104-51-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009463
InChI Key OCKPCBLVNKHBMX-UHFFFAOYSA-N
Synonym n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
PubChem CID 7705
ChEBI CHEBI:44194
IUPAC Name butylbenzene
SMILES CCCCC1=CC=CC=C1
Specifications
CAS Min % 99.0
CAS Max % 100.0
Melting Point -88°C
Color Colorless
Density 0.8600g/mL
Boiling Point 183°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H5(CH2)3CH3
Refractive Index 1.4880 to 1.49
Quantity 50 mL
Beilstein 05,413
Merck Index 15,1551
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble in water. Other solubilities: miscible with ethanol,ether and benzene
Formula Weight 134.22
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Butylbenzene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • n-Butylbenzene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.