missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl propionate, 99+%

Product Code. p-3777788 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10117903

Brand: Thermo Scientific Acros 269482500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-01-2 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-01-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009448
InChI Key BTMVHUNTONAYDX-UHFFFAOYSA-N
Synonym butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
PubChem CID 11529
IUPAC Name butyl propanoate
SMILES CCCCOC(=O)CC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -75°C
Color Colorless
Density 0.8800g/mL
Boiling Point 145°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)3OC(O)C2H5
Refractive Index 1.4000 to 1.402
Quantity 250 mL
Beilstein 02, 241
Merck Index 15, 1590
Specific Gravity 0.88
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.83 mPa.s (20°C)
Formula Weight 130.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Butyl propionate
Show More Show Less
compliance-icons
Product Identifier
  • n-Butyl propionate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.