Specifications
| Melting Point | -90°C |
| Color | Colorless |
| pH | 6.2 |
| Boiling Point | 126°C |
| Packaging | Aluminum can |
| Molecular Formula | C6H12O2 |
| Quantity | 5 L |
| Vapor Density | 4 |
| Formula Weight | 116.16g/mol |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- n-Butyl acetate
- Warning
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.