Learn More
n-Butyl-1,1-d2 Alcohol, CDN
Product Code. 30322373
Change view
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322373 0.25 g 0.25g
30316829 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322373 Supplier CDN Supplier No. CDND66950.25G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Butanol-1,1-D2
CAS 32586-14-4
Molecular Formula C4H8D2O
Quantity 0.25 g
Synonym 1,1-Dideuteriobutan-1-ol
InChI Formula InChI=1 S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i4D2
SMILES OC([2 H])([2 H])CCC
IUPAC Name butan-1,1-d2-1-ol
Molecular Weight (g/mol) 76.13
Formula Weight 76.09 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity