Learn More
N-Boc-L-isoleucine, TRC
Product Code. 30178998
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178998 10 g 10g
30179149 100 g 100g
30179571 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30178998 Supplier Toronto Research Chemicals Supplier No. TRCB65722510G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13139-16-7
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-L-isoleucine, N-Carboxy-L-isoleucine N-tert-Butyl Ester, (2S,3S)-2-(tert-Butoxycarbonylamino)-3-methylpentanoic Acid, (2S,3S)-2-[(tert-Butoxycarbonyl)amino]-3-methylpentanoic Acid, BOC-L-isoleucine, BOC-isoleucine, N-(tert-Butoxycarbonyl)-L-isoleucine, N-(tert-Butyloxycarbonyl)-L-isoleucine, N-Carboxyisoleucine N-tert-Butyl Ester, N-t-Boc-L-isoleucine, N-tert-Butoxycarbonylisoleucine, NSC 334311, tert-Butoxycarbonyl-L-isoleucine
IUPAC Name (2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)O
Specifications
InChI Formula InChI=1S/C11H21NO4/c1-6-7(2)8(9(13)14)12-10(15)16-11(3,4)5/h7-8H,6H2,1-5H3,(H,12,15)(H,13,14)/t7-,8-/m0/s1
Quantity 10 g
Formula Weight 231.1471
Shipping Condition Room Temperature
Chemical Name or Material N-Boc-L-isoleucine