missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-isoleucine, 98+%

Product Code. 11478646
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478646 1 g 1g
11488646 5 g 5g
11498646 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478646 Supplier Thermo Scientific Alfa Aesar Supplier No. A16021.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13139-16-7 | C11H21NO4 | 231.29 g/mol

N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13139-16-7
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00038324 MFCD00237550
InChI Key QJCNLJWUIOIMMF-YUMQZZPRSA-N
Synonym boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine
PubChem CID 2724762
IUPAC Name (2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 67°C to 70°C
Quantity 1 g
Beilstein 1711700
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 231.06
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-isoleucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.