Learn More
N-Boc-glycine ethyl ester, 95%
Product Code. 15419128
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419128 25 g 25g
15409128 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15419128 Supplier Thermo Scientific Alfa Aesar Supplier No. H62611.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 14719-37-0 | C9H17NO4 | 203.238 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14719-37-0
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD05663720
InChI Key CNIBHMMDDXGDNR-UHFFFAOYSA-N
Synonym n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
PubChem CID 357729
IUPAC Name ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CCOC(=O)CNC(=O)OC(C)(C)C
Specifications
Color Colorless
Linear Formula (CH3)3COCONHCH2CO2CH2CH3
Quantity 25 g
Formula Weight 203.24
Percent Purity 95%
Physical Form Oily Liquid
Chemical Name or Material N-Boc-glycine ethyl ester