Learn More
N-Boc-ethylamine, 97%
Product Code. 15415327
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415327 5 g 5g
15405327 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15415327 Supplier Thermo Scientific Alfa Aesar Supplier No. H32575.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 38267-76-4 | C7H15NO2 | 145.202 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38267-76-4
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD12923020
InChI Key ISTGQSQWSKCNFJ-UHFFFAOYSA-N
Synonym tert-butyl ethylcarbamate, n-boc-ethylamine, carbamic acid, ethyl-, 1,1-dimethylethyl ester, acmc-209iz1, ethylcarbamic acid tert-butyl ester
PubChem CID 11198486
IUPAC Name tert-butyl N-ethylcarbamate
SMILES CCNC(=O)OC(C)(C)C
Specifications
Melting Point 38°C to 41°C
Quantity 5 g
Formula Weight 145.2
Percent Purity 97%
Chemical Name or Material N-Boc-ethylamine
compliance-icons
Product Identifier
  • N-Boc-ethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butyl ethylcarbamate