Learn More
N-Boc-D-leucine, TRC
Product Code. 30344164
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344164 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30344164 Supplier Toronto Research Chemicals Supplier No. TRCB65680025G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Boc-D-leucine
CAS 16937-99-8
Molecular Formula C11H21NO4
Quantity 25 g
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-D-leucine, N-Carboxy-D-leucine N-tert-Butyl Ester, (R)-2-(tert-Butoxycarbonylamino)-4-methylpentanoic Acid, D-Boc Leucine, N-(tert-Butoxycarbonyl)-D-leucine, tert-Butoxycarbonyl-D-leucine
InChI Formula InChI=1S/C11H21NO4/c1-7(2)6-8(9(13)14)12-10(15)16-11(3,4)5/h7-8H,6H2,1-5H3,(H,12,15)(H,13,14)/t8-/m1/s1
SMILES CC(C)C[C@@H](NC(=O)OC(C)(C)C)C(=O)O
IUPAC Name (2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
Molecular Weight (g/mol) 231.29
Formula Weight 231.1471