missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-aspartic acid 4-cyclohexyl ester, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
N-Boc-D-aspartic acid 4-cyclohexyl ester is used as pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
N-Boc-D-aspartic acid 4-cyclohexyl ester is used as pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Quantity | 1 g |
IUPAC Name | 2-{[(tert-butoxy)carbonyl]amino}-4-(cyclohexyloxy)-4-oxobutanoic acid |
Formula Weight | 315.36 |
Percent Purity | 95% |
Chemical Name or Material | N-Boc-D-aspartic acid 4-cyclohexyl ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'