Learn More
N-Boc-6-azido-L-norleucine Cyclohexylammonium Salt, TRC
Product Code. 30340319
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340319 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30340319 Supplier Toronto Research Chemicals Supplier No. TRCB624590100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Boc-6-azido-L-norleucine Cyclohexylammonium Salt
CAS 2098497-30-2
Molecular Formula C11H20N4O4 . C6H13N
Quantity 100 mg
Synonym (2S)-6-Azido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic Acid Cyclohexylammonium Salt
InChI Formula InChI=1S/C11H20N4O4.C6H13N/c1-11(2,3)19-10(18)14-8(9(16)17)6-4-5-7-13-15-12;7-6-4-2-1-3-5-6/h8H,4-7H2,1-3H3,(H,14,18)(H,16,17);6H,1-5,7H2/t8-;/m0./s1
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCN=[N+]=[N-])C(=O)O.NC1CCCCC1
IUPAC Name (2S)-6-azido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid;cyclohexanamine
Molecular Weight (g/mol) 272.3
Formula Weight 371.2533