missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-methyl-L-phenylalanine, 95%

Product Code. p-7034312
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488467 250 mg 250mg
15468467 1 g 1g
15478467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488467 Supplier Thermo Scientific Alfa Aesar Supplier No. H51983.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 80102-26-7 | C15H21NO4 | 279.34 g/mol

It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80102-26-7
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD01317027
InChI Key JYRWNPUFECDJCX-LBPRGKRZSA-N
Synonym n-boc-4-methyl-l-phenylalanine, boc-phe 4-me-oh, boc-l-4-methylphe, boc-4-methyl-l-phenylalanine, boc-l-4-methylphenylalanine, boc-4-methy-l-phenylalanine, boc-p-me-phe-oh, boc-l-4-me-phe-oh, 2s-2-tert-butoxycarbonyl amino-3-4-methylphenyl propanoic acid, l-phenylalanine, n-1,1-dimethylethoxy carbonyl-4-methyl
PubChem CID 2761472
SMILES CC1=CC=C(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)C=C1

Specifications

Melting Point 84°C to 88°C
Quantity 250 mg
Beilstein 5381557
Solubility Information Slightly soluble in water.
Optical Rotation +16° (c=1 in Methanol)
IUPAC Name (2S)-3-(4-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Formula Weight 279.34
Percent Purity 95%
Chemical Name or Material N-Boc-4-methyl-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.