Learn More
N-Boc-(3S)-3-phenyl-3-aminopropionaldehyde, TRC
Product Code. 30342571
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342571 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30342571 Supplier Toronto Research Chemicals Supplier No. TRCB6615751MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Boc-(3S)-3-phenyl-3-aminopropionaldehyde
CAS 135865-78-0
Molecular Formula C14 H19 N O3
Quantity 1 mg
Synonym Carbamic acid, N-[(1S)-3-oxo-1-phenylpropyl]-, 1,1-dimethylethyl ester, Carbamic acid, (3-oxo-1-phenylpropyl)-, 1,1-dimethylethyl ester, (S)-, Carbamic acid, [(1S)-3-oxo-1-phenylpropyl]-, 1,1-dimethylethyl ester (9CI), (S)-(3-Oxo-1-phenylpropyl)carbamic acid tert-butyl ester, (S)-3-Phenyl-3-Boc-aminopropionaldehyde, (S)-tert-Butyl (3-oxo-1-phenylpropyl)carbamate, tert-Butyl (1S)-3-oxo-1-phenylpropylcarbamate, N-Boc-(3S)-3-phenyl-3-aminopropionaldehyde, Boc-(S)-3-Amino-3-phenylpropanal, N-Boc-(3S)-3-Amino-3-phenylpropanal
InChI Formula InChI=1S/C14H19NO3/c1-14(2,3)18-13(17)15-12(9-10-16)11-7-5-4-6-8-11/h4-8,10,12H,9H2,1-3H3,(H,15,17)/t12-/m0/s1
SMILES CC(C)(C)OC(=O)N[C@@H](CC=O)c1ccccc1
IUPAC Name tert-butyl N-[(1S)-3-oxo-1-phenylpropyl]carbamate
Molecular Weight (g/mol) 249.31
Formula Weight 249.1365