missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-norleucine, 98+%, Thermo Scientific™

Product Code. 11362786
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362786 1 g 1g
11372786 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362786 Supplier Thermo Scientific Alfa Aesar Supplier No. L08921.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15027-14-2
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00063169
InChI Key NMYWMOZOCYAHNC-GFCCVEGCSA-N
Synonym z-d-norleucine, z-d-nle-oh, n-benzyloxycarbonyl-d-norleucine, r-2-benzyloxy carbonyl amino hexanoic acid, 2r-2-phenylmethoxycarbonylamino hexanoic acid, 2r-2-benzyloxy carbonyl amino hexanoic acid, cbz-d-norleucine, z-d-2-aminohexanoic acid, +-n-benzyloxycarbonyl-d-norleucine, r-2-benzyloxy carbonyl amino hexanoicacid
PubChem CID 7004935
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)hexanoic acid
SMILES CCCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Beilstein 5752565
Optical Rotation +10° (c=1 in Methanol)
Formula Weight 265.3
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-norleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.