missing translation for 'onlineSavingsMsg'
Learn More

N'-Benzyl-N,N-dimethylethylenediamine, 97%, Thermo Scientific™

Product Code. 11382026
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382026 25 g 25g
11372026 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382026 Supplier Thermo Scientific Alfa Aesar Supplier No. L03126.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-55-9
Molecular Formula C11H18N2
Molecular Weight (g/mol) 178.28
MDL Number MFCD00008334
InChI Key LLSJAFHDYCTFCM-UHFFFAOYSA-N
Synonym n'-benzyl-n,n-dimethylethylenediamine, 1,2-ethanediamine, n,n-dimethyl-n'-phenylmethyl, n,n-dimethyl-n'-benzylethylenediamine, benzyl 2-dimethylamino ethyl amine, n'-benzyl-n,n-dimethyl-ethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-n2-phenylmethyl, n'-benzyl-n,n-dimethylethane-1,2-diamine, n'-benzyl-n,n-dimethyl-ethylenediamine, n1-benzyl-n2,n2-dimethyl-1,2-ethanediamine, n1-benzyl-n2,n2-dimethylethane-1,2-diamine
PubChem CID 66029
IUPAC Name N-benzyl-N',N'-dimethylethane-1,2-diamine
SMILES CN(C)CCNCC1=CC=CC=C1

Specifications

Density 0.922
Boiling Point 255°C
Flash Point >110°C (230°F)
Odor Amine-like
Linear Formula C6H5CH2NHCH2CH2N(CH3)2
Refractive Index 1.509
Quantity 25 g
UN Number UN2735
Beilstein 2641339
Formula Weight 178.28
Percent Purity 97%
Chemical Name or Material N'-Benzyl-N,N-dimethylethylenediamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.