Learn More
N'-Benzyl-N,N-dimethylethylenediamine, 97%, Thermo Scientific™
Product Code. 11372026
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372026 5 g 5g
11382026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11372026 Supplier Thermo Scientific Alfa Aesar Supplier No. L03126.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103-55-9
Molecular Formula C11H18N2
Molecular Weight (g/mol) 178.28
MDL Number MFCD00008334
InChI Key LLSJAFHDYCTFCM-UHFFFAOYSA-N
Synonym n'-benzyl-n,n-dimethylethylenediamine, 1,2-ethanediamine, n,n-dimethyl-n'-phenylmethyl, n,n-dimethyl-n'-benzylethylenediamine, benzyl 2-dimethylamino ethyl amine, n'-benzyl-n,n-dimethyl-ethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-n2-phenylmethyl, n'-benzyl-n,n-dimethylethane-1,2-diamine, n'-benzyl-n,n-dimethyl-ethylenediamine, n1-benzyl-n2,n2-dimethyl-1,2-ethanediamine, n1-benzyl-n2,n2-dimethylethane-1,2-diamine
PubChem CID 66029
IUPAC Name N-benzyl-N',N'-dimethylethane-1,2-diamine
SMILES CN(C)CCNCC1=CC=CC=C1
Specifications
Density 0.922
Boiling Point 255°C
Flash Point >110°C (230°F)
Odor Amine-like
Linear Formula C6H5CH2NHCH2CH2N(CH3)2
Refractive Index 1.509
Quantity 5 g
UN Number UN2735
Beilstein 2641339
Formula Weight 178.28
Percent Purity 97%
Chemical Name or Material N'-Benzyl-N,N-dimethylethylenediamine