Learn More
N-Benzyl-4-chlorobenzamide, 97%, Thermo Scientific™
Product Code. p-7036998
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15401808 250 mg 250mg
15491798 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15401808 Supplier Thermo Scientific Alfa Aesar Supplier No. H55517.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7461-34-9
Molecular Formula C14H12ClNO
Molecular Weight (g/mol) 245.71
MDL Number MFCD00018682
InChI Key LSMWDKIFKGLNSW-UHFFFAOYSA-N
Synonym n-benzyl-4-chloro-benzamide, 4-chloro-n-benzylbenzamide, n∼1∼-benzyl-4-chlorobenzamide, 4-chloro-n-phenylmethyl benzamide, 4-chlorophenyl-n-benzylcarboxamide, 4-chloranyl-n-phenylmethyl benzamide
PubChem CID 346668
IUPAC Name N-benzyl-4-chlorobenzamide
SMILES ClC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1
Specifications
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 245.71
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-chlorobenzamide