Learn More
N-Acetylthiourea, TRC
Product Code. 30323124
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323124 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323124 Supplier Toronto Research Chemicals Supplier No. TRCA1888501G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetylthiourea
CAS 591-08-2
Molecular Formula C3H6N2OS
Quantity 1 g
Synonym N-(Aminothioxomethyl)-acetamide, 1-Acetyl-2-thio-urea, 1-Acetyl-2-thiourea, 1-Acetylthiourea, Acetothiourea, Acetylthiocarbamide, Acetylthiourea, N-(Aminothioxomethyl)acetamide, N-Acetyl-2-thiourea, N-Acetylthiocarbamide, NSC 7597
InChI Formula InChI=1S/C3H6N2OS/c1-2(6)5-3(4)7/h1H3,(H3,4,5,6,7)
SMILES CC(=O)NC(=S)N
IUPAC Name N-carbamothioylacetamide
Molecular Weight (g/mol) 118.16
Formula Weight 118.0201