Learn More
N-Acetylthiourea, 99+%, Thermo Scientific™
Product Code. 11434707
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434707 500 g 500g
11414707 25 g 25g
11424707 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11434707 Supplier Thermo Scientific Alfa Aesar Supplier No. B21198.36
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 591-08-2
Molecular Formula C3H6N2OS
Molecular Weight (g/mol) 118.154
MDL Number MFCD00004937
InChI Key IPCRBOOJBPETMF-UHFFFAOYSA-N
Synonym n-acetylthiourea, acetylthiourea, 1-acetylthiourea, 1-acetyl-2-thiourea, acetothiourea, n-acetyl-2-thiourea, acetamide, n-aminothioxomethyl, n-acetylthiocarbamide, urea, 1-acetyl-2-thio, acetylthiocarbamide
PubChem CID 2723593
IUPAC Name N-carbamothioylacetamide
SMILES CC(=O)NC(=S)N
Specifications
Melting Point 165°C to 169°C
Odor Odorless
Quantity 500 g
UN Number UN2811
Beilstein 969960
Formula Weight 118.16
Percent Purity ≥99%
Chemical Name or Material N-Acetylthiourea