Learn More
N-Acetylprocainamide, MedChemExpress
Product Code. 16438616
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16438616 Supplier Medchem Express Supplier No. HYB1109
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress N-Acetylprocainamide is a class III antiarrhythmic, which blocks K+ channels.
N-Acetylprocainamide is a class III antiarrhythmic, which blocks K+ channels.
Specifications
Chemical Name or Material N-Acetylprocainamide
CAS 32795-44-1
Color White
Purity Grade Notes Research
Molecular Formula C15H23N3O2
Quantity 100 mg
Synonym Acecainide NAPA
Solubility Information DMSO : ≥ 100 mg/mL (360.54 mM)
SMILES O=C(NCCN(CC)CC)C1=CC=C(NC(C)=O)C=C1
Molecular Weight (g/mol) 277.36
Formula Weight 277.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation