missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylmannosamine Kinase/GNE Antibody (SR2196), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22587-50ul
This item is not returnable.
View return policy
Description
N-Acetylmannosamine Kinase/GNE Monoclonal antibody specifically detects N-Acetylmannosamine Kinase/GNE in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| N-Acetylmannosamine Kinase/GNE | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR2196 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, DMRV, GLCNENM, glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase, IBM2, N-acylmannosamine kinase, Uae1, UDP-GlcNAc-2-epimerase/ManAc kinase, UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase, UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase | |
| A synthesized peptide derived from human N-Acetylmannosamine Kinase/GNE (Uniprot #: Q9Y223) | |
| 50 μg | |
| Cardiovascular Biology, Cell Biology, Signal Transduction | |
| 10020 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction