Learn More
N-Acetyllactosamine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyllactosamine |
| CAS | 32181-59-2 |
| Molecular Formula | C14 H25 N O11 |
| Quantity | 25 mg |
| Synonym | D-Glucose, 2-(acetylamino)-2-deoxy-4-O-β-D-galactopyranosyl-, D-Glucosamine, N-acetyl-4-O-β-D-galactopyranosyl- (6CI), D-Glucose, 2-acetamido-2-deoxy-4-O-β-D-galactopyranosyl- (7CI,8CI), 2-(Acetylamino)-2-deoxy-4-O-β-D-galactopyranosyl-D-glucose, 2-Acetamido-2-deoxy-4-O-β-D-galactopyranosyl-D-glucose, Lactosamine, N-acetyl-, N-Acetyl-4-O-β-D-galactopyranosyl-D-glucosamine, N-Acetyllactosamine, O-β-D-Galactopyranosyl-(1→4)-2-deoxy-2-acetamido-D-glucose |
| InChI Formula | InChI=1S/C14H25NO11/c1-4(18)15-7-9(20)12(6(3-17)24-13(7)23)26-14-11(22)10(21)8(19)5(2-16)25-14/h5-14,16-17,19-23H,2-3H2,1H3,(H,15,18)/t5-,6-,7-,8+,9-,10+,11-,12-,13?,14+/m1/s1 |
| SMILES | CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@@H]1O |
| IUPAC Name | N-[(3R,4R,5S,6R)-2,4-dihydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-3-yl]acetamide |
| Molecular Weight (g/mol) | 383.35 |
| Formula Weight | 383.1428 |