Learn More
N-Acetyl Sulfamethazine-d4, TRC
Product Code. 30316529
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316529 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316529 Supplier Toronto Research Chemicals Supplier No. TRCA18785210MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetyl Sulfamethazine-d4
CAS 1189891-99-3
Molecular Formula C14H12D4N4O3S
Quantity 10 mg
Synonym N-[4-[[(4,6-Dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl-d4]acetamide, 4'-[(4,6-Dimethyl-2-pyrimidinyl)sulfamoyl]acetanilide-d4, Acetylsulfadimidine-d4, N4-Acetylsulfadimidine-d4, N4-Acetylsulmet-d4
InChI Formula InChI=1S/C14H16N4O3S/c1-9-8-10(2)16-14(15-9)18-22(20,21)13-6-4-12(5-7-13)17-11(3)19/h4-8H,1-3H3,(H,17,19)(H,15,16,18)/i4D,5D,6D,7D
SMILES O=S(NC1=NC(C)=CC(C)=N1)(C2=C([2H])C([2H])=C(NC(C)=O)C([2H])=C2[2H])=O
IUPAC Name N-[2,3,5,6-tetradeuterio-4-[(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
Molecular Weight (g/mol) 324.39