Learn More
N-Acetyl-S-ethyl-L-cysteine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-S-ethyl-L-cysteine |
| CAS | 31386-36-4 |
| Molecular Formula | C7H13NO3S |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C7H13NO3S/c1-3-12-4-6(7(10)11)8-5(2)9/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)/t6-/m0/s1 |
| SMILES | CCSC[C@H](NC(=O)C)C(=O)O |
| IUPAC Name | (2R)-2-acetamido-3-ethylsulfanylpropanoic acid |
| Molecular Weight (g/mol) | 191.24 |
| Formula Weight | 191.0616 |