Learn More
N-Acetyl-N-tert-butoxycarbonyl Serotonin, TRC
Product Code. 30323571
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323571 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323571 Supplier Toronto Research Chemicals Supplier No. TRCA17115510MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetyl-N-tert-butoxycarbonyl Serotonin
CAS 1329837-64-0
Molecular Formula C17H22N2O4
Quantity 10 mg
Synonym N-tert-Butoxycarbonyl-normelatonin, N-tert-Butoxycarbonyl-O-demethylmelatonin, N-[2-(N-tert-Butoxycarbonyl-5-hydroxyindol-3-yl)ethyl]-acetamide, N-tert-Butoxycarbonyl-5-hydroxymelatonin, [1-(N-tert-Butoxycarbonyl)-3-(2-acetamidoethyl)-5-hydroxy]indole
InChI Formula InChI=1S/C17H22N2O4/c1-11(20)18-8-7-12-10-19(16(22)23-17(2,3)4)15-6-5-13(21)9-14(12)15/h5-6,9-10,21H,7-8H2,1-4H3,(H,18,20)
SMILES CC(=O)NCCc1cn(C(=O)OC(C)(C)C)c2ccc(O)cc12
IUPAC Name tert-butyl 3-(2-acetamidoethyl)-5-hydroxyindole-1-carboxylate
Molecular Weight (g/mol) 318.37
Formula Weight 318.158