Learn More
N-Acetyl Mesalazine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyl Mesalazine |
| CAS | 51-59-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 H9 N O4 |
| Quantity | 250 mg |
| Synonym | 5-Acetaminosalicylic Acid |
| InChI Formula | InChI=1S/C9H9NO4/c1-5(11)10-6-2-3-8(12)7(4-6)9(13)14/h2-4,12H,1H3,(H,10,11)(H,13,14) |
| SMILES | CC(=O)Nc1ccc(O)c(c1)C(=O)O |
| IUPAC Name | 5-acetamido-2-hydroxybenzoic acid |
| Molecular Weight (g/mol) | 195.17 |
| Show More |