Learn More
N-Acetyl-L-serine-2,3,3-d3, CDN
Product Code. 30325515
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325515 0.05 g 0.05g
30322497 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325515 Supplier CDN Supplier No. CDND69990.05G
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetyl-L-serine-2,3,3-d3
CAS 2230887-17-7
Molecular Formula C5 D3 H6 N O4
Quantity 0.05 g
Synonym N-Acetyl-L-serine-2,3,3-d3, N-Acetyl-L-serine-α,β,β-D3, N-Acetyl-L-serine-d3, (2 S)-2-Acetamido-3-hydroxypropanoic-2,3,3-d3 acid
InChI Formula InChI=1 S/C5H9NO4/c1-3(8)6-4(2-7)5(9)10/h4,7 H,2H2,1H3,(H,6,8)(H,9,10)/t4-/m0/s1/i2D2,4 D
SMILES [2 H]C([2 H])(O)[C@]([2 H])(NC(=O)C)C(=O)O
IUPAC Name (2 S)-2-acetamido-2,3,3-trideuterio-3-hydroxypropanoic acid
Molecular Weight (g/mol) 150.1478
Formula Weight 150.072 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity