Learn More
N-Acetyl-L-aspartic Acid-d3, TRC
Product Code. 30179569
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179569 10 mg 10mg
30179433 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179569 Supplier Toronto Research Chemicals Supplier No. TRCA16860810MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 284665-15-2
Molecular Formula C6 D3 H6 N O5
Molecular Weight (g/mol) 178.158
Synonym L-Aspartic-2,3,3-d3 acid, N-acetyl- (9CI), (2S)-2-Acetamidobutanedioic-2,3,3-d3 acid, N-Acetyl-L-aspartic-2,3,3-d3 acid, N-Acetyl-L-aspartic-2,3,3-D3 acid
IUPAC Name (2S)-2-acetamido-2,3,3-trideuteriobutanedioic acid
SMILES [2H]C([2H])(C(=O)O)[C@]([2H])(NC(=O)C)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C6H9NO5/c1-3(8)7-4(6(11)12)2-5(9)10/h4H,2H2,1H3,(H,7,8)(H,9,10)(H,11,12)/t4-/m0/s1/i2D2,4D
Formula Weight 178.067
Chemical Name or Material N-Acetyl-L-aspartic Acid-d3